Methyl 3-(3,3-difluorocyclobutyl)-3-oxopropanoate
Lieferant: Apollo Scientific
PC420002-1G
APOSPC420002-1GEA
1340
EUR
APOSPC420002-1G
Methyl 3-(3,3-difluorocyclobutyl)-3-oxopropanoate
Methyl 3-(3,3-difluorocyclobutyl)-3-oxopropanoate
Formel:
C₈H₁₀F₂O₃ Molecular Weight: 192,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD17015923 CAS-Nummer: 1191096-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...