3-Hydroxy-3-(trifluoromethyl)cyclobutanecarboxylic acid

Lieferant: Apollo Scientific

PC420004-1G
APOSPC420004-1GEA 458 EUR
APOSPC420004-1G
3-Hydroxy-3-(trifluoromethyl)cyclobutanecarboxylic acid
3-Hydroxy-3-(trifluoromethyl)cyclobutanecarboxylic acid
Formel: C₆H₇F₃O₃
Molecular Weight: 184,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11870383
CAS-Nummer: 1163729-49-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR