2-Azaspiro[3.3]heptan-6-ol trifluoroacetate

Lieferant: Apollo Scientific

PC420005-1G
APOSPC420005-1GEA 1760 EUR
APOSPC420005-1G
2-Azaspiro[3.3]heptan-6-ol trifluoroacetate
2-Azaspiro[3.3]heptan-6-ol trifluoroacetate
Formel: C₈H₁₂F₃NO₃
Molecular Weight: 227.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23703609
CAS-Nummer: 1256352-97-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR