Diisopropyl-3-fluorocyclobutane-1,1-dicarboxylate

Lieferant: Apollo Scientific

PC420006-1G
APOSPC420006-1GEA 492 EUR
APOSPC420006-1G
Diisopropyl-3-fluorocyclobutane-1,1-dicarboxylate
Diisopropyl-3-fluorocyclobutane-1,1-dicarboxylate
Formel: C₁₂H₁₉FO₄
Molecular Weight: 246,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23105821
CAS-Nummer: 1403767-29-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR