Diisopropyl-3,3-difluorocyclobutane-1,1-dicarboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,1-bis(propan-2-yl) 3,3-difluorocyclobutane-1,1-dicarboxylate

PC420007-1G
APOSPC420007-1GEA 492 EUR
APOSPC420007-1G
Diisopropyl-3,3-difluorocyclobutane-1,1-dicarboxylate
Diisopropyl-3,3-difluorocyclobutane-1,1-dicarboxylate
Formel: C₁₂H₁₈F₂O₄
Molecular Weight: 264,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23105827
CAS-Nummer: 1225532-89-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR