3,3-Difluorocyclobutane-1,1-dicarboxylic-1-isopropyl ester

Lieferant: Apollo Scientific
Achtung

PC420008-1G
APOSPC420008-1GEA 690 EUR
APOSPC420008-1G
3,3-Difluorocyclobutane-1,1-dicarboxylic-1-isopropyl ester
3,3-Difluorocyclobutane-1,1-dicarboxylic-1-isopropyl ester
Formel: C₉H₁₂F₂O₄
Molecular Weight: 222,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23105828
CAS-Nummer: 1225532-90-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR