1-(Boc-amino)-3,3-difluorocyclobutanecarboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-((tert-Butoxycarbonyl)amino)-3,3-difluorocyclobutanecarboxylic acid
PC420009-1G
APOSPC420009-1GEA
1100
EUR
APOSPC420009-1G
1-(Boc-amino)-3,3-difluorocyclobutanecarboxylic acid
1-(Boc-amino)-3,3-difluorocyclobutanecarboxylic acid
Formel:
C₁₀H₁₅F₂NO₄ Molecular Weight: 251,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22415254 CAS-Nummer: 1363380-83-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...