1-Amino-3,3-difluorocyclobutane-1-methanol hydrochloride

Lieferant: Apollo Scientific

PC420010-1G
APOSPC420010-1GEA 1250 EUR
APOSPC420010-1G
1-Amino-3,3-difluorocyclobutane-1-methanol hydrochloride
1-Amino-3,3-difluorocyclobutane-1-methanol hydrochloride
Formel: C₅H₁₀ClF₂NO
Molecular Weight: 173.59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23703610
CAS-Nummer: 1363380-82-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR