3,3-Difluoro-1-methylcyclobutanecarboxylic acid

Lieferant: Apollo Scientific

PC420011-1G
APOSPC420011-1GEA 1080 EUR
APOSPC420011-1G
3,3-Difluoro-1-methylcyclobutanecarboxylic acid
3,3-Difluoro-1-methylcyclobutanecarboxylic acid
Formel: C₆H₈F₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD20645342
CAS-Nummer: 227607-43-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR