3,3-Difluoro-1-methylcyclobutanamine hydrochloride

Lieferant: Apollo Scientific
Achtung

PC420012-1G
APOSPC420012-1GEA 1730 EUR
APOSPC420012-1G
3,3-Difluoro-1-methylcyclobutanamine hydrochloride
3,3-Difluoro-1-methylcyclobutanamine hydrochloride
Formel: C₅H₉F₂N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD26959150
CAS-Nummer: 1523606-39-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR