3-Amino-1-(trifluoromethyl)cyclobutan-1-ol hydrochloride

Lieferant: Apollo Scientific

PC420014-1G
APOSPC420014-1GEA 1140 EUR
APOSPC420014-1G
3-Amino-1-(trifluoromethyl)cyclobutan-1-ol hydrochloride
3-Amino-1-(trifluoromethyl)cyclobutan-1-ol hydrochloride
Formel: C₅H₉ClF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD22392108
CAS-Nummer: 1408075-12-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR