(3,3-Difluorocyclobutyl)-N-methylmethanamine hydrochloride
Lieferant: Apollo Scientific
PC420015-1G
APOSPC420015-1GEA
1140
EUR
APOSPC420015-1G
(3,3-Difluorocyclobutyl)-N-methylmethanamine hydrochloride
(3,3-Difluorocyclobutyl)-N-methylmethanamine hydrochloride
Formel:
C₆H₁₂ClF₂N Lagertemperatur: Raumtemperatur |
MDL:
MFCD16987777 CAS-Nummer: 1250444-03-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...