3-Azetidineacetic acid trifluoroacetate 95%
Lieferant: Apollo Scientific
PC420016-1G
APOSPC420016-1GEA
183
EUR
APOSPC420016-1G
3-Azetidineacetic acid trifluoroacetate 95%
3-Azetidineacetic acid trifluoroacetate
Formel:
C₅H₉NO₂·CF₃CO₂H Molecular Weight: 229.154 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27986865 CAS-Nummer: 1202076-02-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...