Methyl-3-azetidineacetate trifluoroacetate salt
Lieferant: Apollo Scientific
Synonyme:
Methyl 2-(azetidin-3-yl)acetate 2,2,2-trifluoroacetate
PC420017-1G
APOSPC420017-1GEA
585
EUR
APOSPC420017-1G
Methyl-3-azetidineacetate trifluoroacetate salt
Methyl-3-azetidineacetate trifluoroacetate salt
Formel:
C₈H₁₂F₃NO₄ Molecular Weight: 243,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18425634 CAS-Nummer: 1313738-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...