3-Fluoro-1,3-azetidinedicarboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

Synonyme: 1-(tert-Butoxycarbonyl)-3-fluoroazetidine-3-carboxylic acid

PC420018-1G
APOSPC420018-1GEA 227 EUR
APOSPC420018-1G
3-Fluoro-1,3-azetidinedicarboxylic acid tert-butyl ester
3-Fluoro-1,3-azetidinedicarboxylic acid tert-butyl ester
Formel: C₉H₁₄FNO₄
Molecular Weight: 219,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15071784
CAS-Nummer: 1126650-67-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR