3,3-Difluoro-1-(diphenylmethyl)azetidine

Lieferant: Apollo Scientific

PC420019-5G
APOSPC420019-5GEA 371 EUR
APOSPC420019-5G
3,3-Difluoro-1-(diphenylmethyl)azetidine
3,3-Difluoro-1-(diphenylmethyl)azetidine
Formel: C₁₆H₁₅F₂N
Molecular Weight: 259.294 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09264457
CAS-Nummer: 288315-02-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR