Methyl-1-boc-3-fluoroazetidine-3-carboxylate

Lieferant: Apollo Scientific

PC420020-1G
APOSPC420020-1GEA 1680 EUR
APOSPC420020-1G
Methyl-1-boc-3-fluoroazetidine-3-carboxylate
Methyl-1-boc-3-fluoroazetidine-3-carboxylate
Formel: C₁₀H₁₆FNO₄
Molecular Weight: 233,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20922778
CAS-Nummer: 1363382-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR