1-Boc-3-fluoroazetidine-3-methanol

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 3-fluoro-3-(hydroxymethyl)azetidine-1-carboxylate

PC420021-1G
APOSPC420021-1GEA 212 EUR
APOSPC420021-1G
1-Boc-3-fluoroazetidine-3-methanol
1-Boc-3-fluoroazetidine-3-methanol
Formel: C₉H₁₆FNO₃
Molecular Weight: 205,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17392630
CAS-Nummer: 1126650-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR