3-(Trifluoromethyl)azetidin-3-ol hydrochloride

Lieferant: Apollo Scientific

Synonyme: 3-Trifluoromethyl-3-azetidinol hydrochloride

PC420022-1G
APOSPC420022-1GEA 884 EUR
APOSPC420022-1G
3-(Trifluoromethyl)azetidin-3-ol hydrochloride
3-(Trifluoromethyl)azetidin-3-ol hydrochloride
Formel: C₄H₇ClF₃NO
Molecular Weight: 177,551 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13248564
CAS-Nummer: 848192-96-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR