3-Fluoro-3-methylazetidine hydrochloride
Lieferant: Apollo Scientific
PC420023-1G
APOSPC420023-1GEA
1760
EUR
APOSPC420023-1G
3-Fluoro-3-methylazetidine hydrochloride
3-Fluoro-3-methylazetidine hydrochloride
Formel:
C₄H₉ClFN Lagertemperatur: Raumtemperatur |
MDL:
MFCD21605303 CAS-Nummer: 1427379-42-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...