1-(Perfluoro-n-hexyl)tetradecane

Lieferant: Apollo Scientific
Achtung

PC4208-1G PC4208-5G
APOSPC4208-1GEA 139 EUR
APOSPC4208-1G APOSPC4208-5G
1-(Perfluoro-n-hexyl)tetradecane
1-(Perfluoro-n-hexyl)tetradecane
Formel: C₂₀H₂₉F₁₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04038341
CAS-Nummer: 154628-00-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR