6-Chloro-2,3-difluoro-4-(trifluoromethyl)phenylhydrazine 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Chloro-2,3-difluoro-4-hydrazinobenzotrifluoride
PC4209-1G
APOSPC4209-1GEA
551
EUR
APOSPC4209-1G
6-Chloro-2,3-difluoro-4-(trifluoromethyl)phenylhydrazine 98%
6-Chloro-2,3-difluoro-4-(trifluoromethyl)phenylhydrazine
Formel:
C₇H₄ClF₅N₂ Molecular Weight: 246,565 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD04972718 CAS-Nummer: 120769-98-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...