2-(3,4-Difluorophenyl)-4'-methoxyacetophenone

Lieferant: Apollo Scientific

PC421002-1G
APOSPC421002-1GEA 321 EUR
APOSPC421002-1G
2-(3,4-Difluorophenyl)-4'-methoxyacetophenone
2-(3,4-Difluorophenyl)-4'-methoxyacetophenone
Formel: C₁₅H₁₂F₂O₂
Molecular Weight: 262,251 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12974696
CAS-Nummer: 221179-71-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR