2-(3,4-Difluorophenyl)-2'-fluoroacetophenone
Lieferant: Apollo Scientific
PC421003-1G
APOSPC421003-1GEA
364
EUR
APOSPC421003-1G
2-(3,4-Difluorophenyl)-2'-fluoroacetophenone
2-(3,4-Difluorophenyl)-2'-fluoroacetophenone
Formel:
C₁₄H₉F₃O Molecular Weight: 250,216 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24969058 CAS-Nummer: 1519952-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...