2-(3,4-Difluorophenyl)-3'-fluoroacetophenone

Lieferant: Apollo Scientific

PC421004-1G
APOSPC421004-1GEA 364 EUR
APOSPC421004-1G
2-(3,4-Difluorophenyl)-3'-fluoroacetophenone
2-(3,4-Difluorophenyl)-3'-fluoroacetophenone
Formel: C₁₄H₉F₃O
Molecular Weight: 250,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24829746
CAS-Nummer: 1517619-41-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR