2-(3,4-Difluorophenyl)-4'-methylacetophenone
Lieferant: Apollo Scientific
PC421005-1G
APOSPC421005-1GEA
364
EUR
APOSPC421005-1G
2-(3,4-Difluorophenyl)-4'-methylacetophenone
2-(3,4-Difluorophenyl)-4'-methylacetophenone
Formel:
C₁₅H₁₂F₂O Molecular Weight: 246,252 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23417260 CAS-Nummer: 1504609-61-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...