2-(3-Fluorophenyl)-4'-methoxyacetophenone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(3-Fluorophenyl)-1-(4-methoxyphenyl)ethanone
PC421008-1G
APOSPC421008-1GEA
306
EUR
APOSPC421008-1G
2-(3-Fluorophenyl)-4'-methoxyacetophenone
2-(3-Fluorophenyl)-4'-methoxyacetophenone
Formel:
C₁₅H₁₃FO₂ Schmelzpunkt: 73…75 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11545169 CAS-Nummer: 153529-18-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...