2-(3-Fluorophenyl)-4'-methylacetophenone

Lieferant: Apollo Scientific

PC421009-1G
APOSPC421009-1GEA 611 EUR
APOSPC421009-1G
2-(3-Fluorophenyl)-4'-methylacetophenone
2-(3-Fluorophenyl)-4'-methylacetophenone
Formel: C₁₅H₁₃FO
Lagertemperatur: Raumtemperatur
MDL: MFCD11105606
CAS-Nummer: 1152812-07-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR