2-(3-Fluorophenyl)-4'-fluoroacetophenone

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(3-Fluorophenyl)-1-(4-fluorophenyl)ethanone

PC421012-1G
APOSPC421012-1GEA 306 EUR
APOSPC421012-1G
2-(3-Fluorophenyl)-4'-fluoroacetophenone
2-(3-Fluorophenyl)-4'-fluoroacetophenone
Formel: C₁₄H₁₀F₂O
Molecular Weight: 232,225 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11210358
CAS-Nummer: 370874-66-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR