4'-Fluoro-2-(4-fluorophenyl)acetophenone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,2-Bis(4-fluorophenyl)ethanone
PC421013-1G
APOSPC421013-1GEA
221
EUR
APOSPC421013-1G
4'-Fluoro-2-(4-fluorophenyl)acetophenone
4'-Fluoro-2-(4-fluorophenyl)acetophenone
Formel:
C₁₄H₁₀F₂O Molecular Weight: 232,225 g/mol Siedepunkt: 178…180 °C (15 mm Hg) Schmelzpunkt: 94…98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00055468 CAS-Nummer: 366-68-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...