2-(2,4-Dichlorophenyl)-3'-fluoroacetophenone

Lieferant: Apollo Scientific

PC421015-1G
APOSPC421015-1GEA 350 EUR
APOSPC421015-1G
2-(2,4-Dichlorophenyl)-3'-fluoroacetophenone
2-(2,4-Dichlorophenyl)-3'-fluoroacetophenone
Formel: C₁₄H₉Cl₂FO
Lagertemperatur: Raumtemperatur
MDL: MFCD12723565
CAS-Nummer: 1179892-14-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR