2-(4-Chlorophenyl)-2'-fluoroacetophenone

Lieferant: Apollo Scientific

PC421016-1G
APOSPC421016-1GEA 306 EUR
APOSPC421016-1G
2-(4-Chlorophenyl)-2'-fluoroacetophenone
2-(4-Chlorophenyl)-2'-fluoroacetophenone
Formel: C₁₄H₁₀ClFO
Molecular Weight: 248,68 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16350637
CAS-Nummer: 1183199-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR