2-(4-Chlorophenyl)-3'-fluoroacetophenone

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-Chlorophenyl)-1-(3-fluorophenyl)ethanone

PC421017-1G
APOSPC421017-1GEA 166 EUR
APOSPC421017-1G
2-(4-Chlorophenyl)-3'-fluoroacetophenone
2-(4-Chlorophenyl)-3'-fluoroacetophenone
Formel: C₁₄H₁₀ClFO
Molecular Weight: 248,68 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12605188
CAS-Nummer: 1183554-70-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR