ω-(4-Fluorphenyl)acetophenon
Lieferant: Apollo Scientific
|
Achtung
|
PC421018-1G
APOSPC421018-1GEA
281
EUR
APOSPC421018-1G
ω-(4-Fluorphenyl)acetophenon
ω-(4-Fluorphenyl)acetophenon
Formel:
C₁₄H₁₁FO Schmelzpunkt: 108…111 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02260684 CAS-Nummer: 347-91-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...