4-(Trifluoromethoxy)-2-(trifluoromethyl)aniline hydrochloride

Lieferant: Apollo Scientific
Achtung

PC421019-1G PC421019-250MG
APOSPC421019-1GEA 0 EUR
APOSPC421019-1G APOSPC421019-250MG
4-(Trifluoromethoxy)-2-(trifluoromethyl)aniline hydrochloride
4-(Trifluoromethoxy)-2-(trifluoromethyl)aniline hydrochloride
Formel: C₈H₅F₆NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD27923427
CAS-Nummer: 1588441-12-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR