6-Chloro-3-iodo-2-(trifluoromethoxy)pyridine

Lieferant: Apollo Scientific
Achtung

PC421024-1G
APOSPC421024-1GEA 1450 EUR
APOSPC421024-1G
6-Chloro-3-iodo-2-(trifluoromethoxy)pyridine
6-Chloro-3-iodo-2-(trifluoromethoxy)pyridine
Formel: C₆H₂ClF₃INO
Molecular Weight: 323,439 g/mol
Schmelzpunkt: 33…35 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD22571777
CAS-Nummer: 1221171-95-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR