Methyl 4-chloro-3-fluoropicolinate 98%
Lieferant: Apollo Scientific
PC421039-250MG
APOSPC421039-250MGEA
49.3
EUR
APOSPC421039-250MG
Methyl 4-chloro-3-fluoropicolinate 98%
Methyl 4-chloro-3-fluoropicolinate
Formel:
C₇H₅ClFNO₂ Molecular Weight: 189,572 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16293406 CAS-Nummer: 1034921-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...