2-Chloro-4-trifluoromethoxy-benzaldehyde 95%

Lieferant: Apollo Scientific

PC421043-1G PC421043-250MG PC421043-5G
APOSPC421043-1GEA 224 EUR
APOSPC421043-1G APOSPC421043-250MG APOSPC421043-5G
2-Chloro-4-trifluoromethoxy-benzaldehyde 95%
2-Chloro-4-trifluoromethoxy-benzaldehyde
Formel: C₈H₄ClF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18393768
CAS-Nummer: 1079351-20-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR