2,4-Difluoro-1-(2-fluorophenyl)benzene 95%

Lieferant: Apollo Scientific

Synonyme: 2,2',4-Trifluoro-1,1'-biphenyl

PC421049-250MG
APOSPC421049-250MGEA 231 EUR
APOSPC421049-250MG
2,4-Difluoro-1-(2-fluorophenyl)benzene 95%
2,4-Difluoro-1-(2-fluorophenyl)benzene
Formel: C₁₂H₇F₃
Lagertemperatur: Raumtemperatur
MDL: MFCD22051120
CAS-Nummer: 115245-05-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR