5-Iodo-6-(trifluoromethyl)pyridin-2-ol 95+%

Lieferant: Apollo Scientific

PC421074-250MG
APOSPC421074-250MGEA 30.6 EUR
APOSPC421074-250MG
5-Iodo-6-(trifluoromethyl)pyridin-2-ol 95+%
5-Iodo-6-(trifluoromethyl)pyridin-2-ol
Formel: C₆H₃F₃INO
Molecular Weight: 288,994 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16611365
CAS-Nummer: 1227515-14-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR