6-Bromo-8-fluoro-3,4-dihydroisoquinolin-1(2h)-one 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Bromo-8-fluoro-3,4-dihydroisoquinolin-1(2H)-one
PC421086-250MG
APOSPC421086-250MGEA
235
EUR
APOSPC421086-250MG
6-Bromo-8-fluoro-3,4-dihydroisoquinolin-1(2h)-one 95%
6-Bromo-8-fluoro-3,4-dihydroisoquinolin-1(2h)-one
Formel:
C₉H₇BrFNO Molecular Weight: 244,06 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22381969 CAS-Nummer: 1242157-15-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...