2-(Trifluoromethyl)thiazol-5-amine

Lieferant: Apollo Scientific
Achtung

PC421096-250MG
APOSPC421096-250MGEA 185 EUR
APOSPC421096-250MG
2-(Trifluoromethyl)thiazol-5-amine
2-(Trifluoromethyl)thiazol-5-amine
Formel: C₄H₃F₃N₂S
Molecular Weight: 168,14 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11847091
CAS-Nummer: 1367944-72-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR