1-Methyl-1-(2-nitro-4-(trifluormethyl)phenyl)hydrazin 97%

Lieferant: Apollo Scientific
Achtung

PC4211-1G
APOSPC4211-1GEA 0 EUR
APOSPC4211-1G
1-Methyl-1-(2-nitro-4-(trifluormethyl)phenyl)hydrazin 97%
1-Methyl-1-(2-nitro-4-(trifluoromethyl)phenyl)hydrazine
Formel: C₈H₈F₃N₃O₂
Schmelzpunkt: 64…67 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052691
CAS-Nummer: 120940-43-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR