2-Bromo-5-(trifluoromethyl)thiophene 95+%

Lieferant: Apollo Scientific

PC421102-250MG
APOSPC421102-250MGEA 174 EUR
APOSPC421102-250MG
2-Bromo-5-(trifluoromethyl)thiophene 95+%
2-Bromo-5-(trifluoromethyl)thiophene
Formel: C₅H₂BrF₃S
Molecular Weight: 231,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08461914
CAS-Nummer: 143469-22-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR