5-Bromo-6-fluoropyridin-3-ol 95+%

Lieferant: Apollo Scientific

PC421126-250MG
APOSPC421126-250MGEA 173 EUR
APOSPC421126-250MG
5-Bromo-6-fluoropyridin-3-ol 95+%
5-Bromo-6-fluoropyridin-3-ol
Formel: C₅H₃BrFNO
Molecular Weight: 191,986 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08061303
CAS-Nummer: 186593-54-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR