5-Methyl-2-(trifluoromethyl)pyridin-4-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-methyl-2-(trifluoromethyl)-4-pyridinamine

PC421137-250MG
APOSPC421137-250MGEA 799 EUR
APOSPC421137-250MG
5-Methyl-2-(trifluoromethyl)pyridin-4-amine
5-Methyl-2-(trifluoromethyl)pyridin-4-amine
Formel: C₇H₇F₃N₂
Molecular Weight: 176,139 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18258830
CAS-Nummer: 340809-48-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR