5-(Trifluoromethyl)pyrimidin-2-amine 97%

Lieferant: Apollo Scientific

PC421149-250MG
APOSPC421149-250MGEA 115 EUR
APOSPC421149-250MG
5-(Trifluoromethyl)pyrimidin-2-amine 97%
5-(Trifluoromethyl)pyrimidin-2-amine
Formel: C₅H₄F₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD10696951
CAS-Nummer: 69034-08-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR