2-Fluoro-5-iodopyrimidine 95+%

Lieferant: Apollo Scientific

PC421150-250MG
APOSPC421150-250MGEA 59.6 EUR
APOSPC421150-250MG
2-Fluoro-5-iodopyrimidine 95+%
2-Fluoro-5-iodopyrimidine
Formel: C₄H₂FIN₂
Molecular Weight: 223,975 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09038025
CAS-Nummer: 697300-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR