2-Chloro-3-fluoro-5-(trifluoromethyl)pyridine 98%

Lieferant: Apollo Scientific
Achtung

PC421151-250MG
APOSPC421151-250MGEA 63.8 EUR
APOSPC421151-250MG
2-Chloro-3-fluoro-5-(trifluoromethyl)pyridine 98%
2-Chloro-3-fluoro-5-(trifluoromethyl)pyridine
Formel: C₆H₂ClF₄N
Molecular Weight: 199,533 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185448
CAS-Nummer: 72600-67-0
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR