6-Chloro-5-(trifluoromethyl)pyridin-2-amine 98%

Lieferant: Apollo Scientific

PC421157-1G
APOSPC421157-1GEA 108 EUR
APOSPC421157-1G
6-Chloro-5-(trifluoromethyl)pyridin-2-amine 98%
6-Chloro-5-(trifluoromethyl)pyridin-2-amine
Formel: C₆H₄ClF₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11848573
CAS-Nummer: 79456-28-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR